Learn More
Medchemexpress LLC IR415 25mg | 452967-14-5 | 296.34 | C13H14F2N4S | 25 MG
SDP
Catalog No. 5000228970
Encompass_Preferred
IR415 is a research compound that inhibits hepatitis B virus (HBV) replication by blocking HBx activity. It selectively binds HBx (reported Kd = 2 nM) and reverses HBx-mediated suppression of RNA interference; characterized for dose-dependent inhibition of HBx in vitro.
  • Selective HBx inhibitor (Kd = 2 nM).
  • Demonstrated dose-dependent inhibition of HBx activity in cell assays.
  • Chemical formula C13H14F2N4S; molecular weight 296.34.
  • High purity (reported ≥98% by HPLC).
  • Available as a powdered research reagent; store powder at -20°C for long-term stability.
Catalog No. 50-002-28970 Supplier Medchemexpress LLC Supplier No. HY11699925MG
IR415 is a research compound that inhibits hepatitis B virus (HBV) replication by blocking HBx activity. It selectively binds HBx (reported Kd = 2 nM) and reverses HBx-mediated suppression of RNA interference; characterized for dose-dependent inhibition of HBx in vitro.
  • Selective HBx inhibitor (Kd = 2 nM).
  • Demonstrated dose-dependent inhibition of HBx activity in cell assays.
  • Chemical formula C13H14F2N4S; molecular weight 296.34.
  • High purity (reported ≥98% by HPLC).
  • Available as a powdered research reagent; store powder at -20°C for long-term stability.