Learn More
Medchemexpress LLC IR415 50mg | 452967-14-5 | 50 MG
SDP
Catalog No. 5000229883
Encompass_Preferred
IR415 is a small-molecule research compound that inhibits hepatitis B virus (HBV) replication by blocking HBx activity. It is supplied as a solid (white to off-white) for preclinical research.
  • Potent anti-HBV activity via HBx inhibition.
  • High purity: 99.66% (typical).
  • Solid form, white to off-white appearance.
  • Molecular formula C13H14F2N4S; molecular weight 296.34.
  • Provided in 50 MG research quantities suitable for preclinical use.
Catalog No. 50-002-29883 Supplier Medchemexpress LLC Supplier No. HY11699950MG
IR415 is a small-molecule research compound that inhibits hepatitis B virus (HBV) replication by blocking HBx activity. It is supplied as a solid (white to off-white) for preclinical research.
  • Potent anti-HBV activity via HBx inhibition.
  • High purity: 99.66% (typical).
  • Solid form, white to off-white appearance.
  • Molecular formula C13H14F2N4S; molecular weight 296.34.
  • Provided in 50 MG research quantities suitable for preclinical use.