Learn More
Selleck Chemical LLC IRAK-1-4 Inhibitor I
SDP
Catalog No. 5000002780
Encompass_Preferred
IRAK-1-4 Inhibitor I is an inhibitor of interleukin-1 receptor associated kinase (IRAK) with IC50 values of 0 3 M and 0 2 M for IRAK-1 and IRAK-4 respectively It displays IC50 values 10 M in a panel of 27 other kinases tested
Catalog No. 50-000-02780 Supplier Selleck Chemical LLC Supplier No. S659825MG
Add to Cart
Add to Cart
IRAK-1-4 Inhibitor I is an inhibitor of interleukin-1 receptor associated kinase (IRAK) with IC50 values of 0 3 M and 0 2 M for IRAK-1 and IRAK-4 respectively It displays IC50 values 10 M in a panel of 27 other kinases tested