Learn More
Medchemexpress LLC HY-13275 5mg Medchemexpress, IRAK inhibitor 1 CAS:1042224-63-4 Purity:>98%
SDP
Catalog No. 502081322
Encompass_Preferred
Medchemexpress, HY-13275 5mg IRAK inhibitor 1 CAS:1042224-63-4 IRAK inhibitor 1 is a potent IRAK-4 inhibitor with IC50 of 216 nM, is poorly active against JNK-1 and JNK-2 with IC50 of 3.801 μM, and >10 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1322 Supplier Medchemexpress LLC Supplier No. HY132755MG
Medchemexpress, HY-13275 5mg IRAK inhibitor 1 CAS:1042224-63-4 IRAK inhibitor 1 is a potent IRAK-4 inhibitor with IC50 of 216 nM, is poorly active against JNK-1 and JNK-2 with IC50 of 3.801 μM, and >10 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.