Learn More
Medchemexpress LLC HY-13280 5mg Medchemexpress, IRAK inhibitor 6 CAS:1042672-97-8 Purity:>98%
SDP
Catalog No. 501932659
Encompass_Preferred
Medchemexpress, HY-13280 5mg IRAK inhibitor 6 CAS:1042672-97-8 IRAK inhibitor 6 is an inhibitor of interleukin-1 receptor associated kinase 4 (IRAK-4) with IC 50 of 160 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2659 Supplier Medchemexpress LLC Supplier No. HY132805MG
Medchemexpress, HY-13280 5mg IRAK inhibitor 6 CAS:1042672-97-8 IRAK inhibitor 6 is an inhibitor of interleukin-1 receptor associated kinase 4 (IRAK-4) with IC 50 of 160 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.