Learn More
Cayman Chemical IRAK InhIbItr 1 1mg
SDP
Catalog No. 502913601
Encompass_Preferred
An IRAK4 inhibitor (IC50 = 35 nM); selective for IRAK4 over JNK1 and JNK2 at 10 µM; prevents increases in TNF-α and IL-12p40 levels in M. bovis-infected THP-1 cells at 1 µM
Catalog No. 50-291-3601 Supplier Cayman Chemical Supplier No. 366371 MG
Add to Cart
Add to Cart
An IRAK4 inhibitor (IC50 = 35 nM); selective for IRAK4 over JNK1 and JNK2 at 10 µM; prevents increases in TNF-α and IL-12p40 levels in M. bovis-infected THP-1 cells at 1 µM