Learn More
Cayman Chemical IRAK Inhibitor 1, Purity: ≥98%, CAS: 1042224-63-4, 5 mg
SDP
Catalog No. 502909305
Encompass_Preferred
An IRAK4 inhibitor (IC50 = 35 nM); selective for IRAK4 over JNK1 and JNK2 at 10 µM; prevents increases in TNF-α and IL-12p40 levels in M. bovis-infected THP-1 cells at 1 µM
Catalog No. 50-290-9305 Supplier Cayman Chemical Supplier No. 366375 MG
An IRAK4 inhibitor (IC50 = 35 nM); selective for IRAK4 over JNK1 and JNK2 at 10 µM; prevents increases in TNF-α and IL-12p40 levels in M. bovis-infected THP-1 cells at 1 µM