Learn More
Cayman Chemical IRAK InhIbItr 6 10mg
SDP
Catalog No. 502949115
Encompass_Preferred
An IRAK4 inhibitor (IC50 = 160 nM)
Catalog No. 50-294-9115 Supplier Cayman Chemical Supplier No. 3326710 MG
An IRAK4 inhibitor (IC50 = 160 nM)