Learn More
Cayman Chemical IRAK Inhibitor 6, Purity: ≥98%, CAS: 1042672-97-8, 5 mg
SDP
Catalog No. 502947773
Encompass_Preferred
An IRAK4 inhibitor (IC50 = 160 nM)
Catalog No. 50-294-7773 Supplier Cayman Chemical Supplier No. 332675 MG
An IRAK4 inhibitor (IC50 = 160 nM)