Learn More
Eurofins Discoverx IRAK1, active, 250 µg. (14-684M)
Supplier:  Eurofins Discoverx 14684M
Encompass
N-terminal 6xHis-tagged recombinant, human IRAK1 amino acids 194-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9184