Learn More
Enzo Life Sciences IRAK1 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905709100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human IRAK-1. UniProt ID: P51617. Species reactivity: Human, Mouse, Rat. Applications: ICC, IP, WB. Purity: DEAE purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-7002