Learn More
Cayman Chemical IRAK14 InhIbItr 10mg
SDP
Catalog No. 502964878
Encompass_Preferred
A benzimidazole that selectively disrupts the activity of IRAK 1 and 4 (IC50 = 0.3 and 0.2 µM, respectively)
Catalog No. 50-296-4878 Supplier Cayman Chemical Supplier No. 1754010 MG
A benzimidazole that selectively disrupts the activity of IRAK 1 and 4 (IC50 = 0.3 and 0.2 µM, respectively)