Learn More
Cayman Chemical IRAK1/4 Inhibitor, Purity: ≥98%, CAS: 509093-47-4, 25 mg
SDP
Catalog No. 502966624
Encompass_Preferred
A benzimidazole that selectively disrupts the activity of IRAK 1 and 4 (IC50 = 0.3 and 0.2 µM, respectively)
Catalog No. 50-296-6624 Supplier Cayman Chemical Supplier No. 1754025 MG
A benzimidazole that selectively disrupts the activity of IRAK 1 and 4 (IC50 = 0.3 and 0.2 µM, respectively)