Learn More
Aviva Systems Biology Corp IRAK2 Antibody - C-terminal region (ARP61222_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61222_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IRAK2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human IRAK2. Alternate names: IRAK-2, MGC150550ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-287-301