Learn More
Enzo Life Sciences IRAK 4 polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPST206E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human IRAK-4. UniProt: Q9NWZ3. Species Reactivity: Human. Applications: ICC, WB. Recommended dilutions: Immunocytochemistry (10µg/ml). Western Blot (1-2µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-666-593