Learn More
Medchemexpress LLC IRAK4 Human sf9 H 50ug
SDP
Catalog No. 5000181217
Encompass_Preferred
The IRAK4 protein is a serine/threonine protein kinase that is critical for initiating the innate immune response through the Toll-like receptor (TLR) and IL-1R signaling pathways After TLR is activated it rapidly forms the Myddosome with IRAK2 IRAK4 Protein Human (sf9 His) is the recombinant human-derived IRAK4 protein expressed by Sf9 insect cells with N-10 His labeled tag
Catalog No. 50-001-81217 Supplier Medchemexpress LLC Supplier No. HYP7325550UG
The IRAK4 protein is a serine/threonine protein kinase that is critical for initiating the innate immune response through the Toll-like receptor (TLR) and IL-1R signaling pathways After TLR is activated it rapidly forms the Myddosome with IRAK2 IRAK4 Protein Human (sf9 His) is the recombinant human-derived IRAK4 protein expressed by Sf9 insect cells with N-10 His labeled tag