Learn More
Selleck Chemical LLC IRAK4-IN-1 S0008-5MG
SDP
Catalog No. 5000040247
Encompass_Preferred
IRAK4-IN-1 is an interleukin-1 receptor associated kinase 4 (IRAK4) inhibitor with IC50 of 7 nM
Catalog No. 50-000-40247 Supplier Selleck Chemical LLC Supplier No. S00085MG
IRAK4-IN-1 is an interleukin-1 receptor associated kinase 4 (IRAK4) inhibitor with IC50 of 7 nM