Learn More
Medchemexpress LLC IRAK4-IN-4 100mg | 1850276-58-2 | 100 MG
SDP
Catalog No. 5000227062
Encompass_Preferred
IRAK4-IN-4 is a small-molecule research inhibitor of interleukin-1 receptor-associated kinase 4 (IRAK4) with low-nanomolar potency. It is supplied as a light yellow solid, with molecular formula C22H16N2O2, molecular weight 340.37 g/mol, and high analytical purity. Solubility and storage guidance are provided for in vitro and in vivo use.
  • Low-nanomolar IRAK4 potency (IC50 2.8 nM).
  • Also inhibits cGAS (IC50 2.1 nM).
  • High purity (99.55%).
  • Soluble in DMSO (25 mg/mL); in vivo formulation protocols provided.
  • Light yellow solid with recommended storage conditions for powder and solutions.
Catalog No. 50-002-27062 Supplier Medchemexpress LLC Supplier No. HY114181100MG
IRAK4-IN-4 is a small-molecule research inhibitor of interleukin-1 receptor-associated kinase 4 (IRAK4) with low-nanomolar potency. It is supplied as a light yellow solid, with molecular formula C22H16N2O2, molecular weight 340.37 g/mol, and high analytical purity. Solubility and storage guidance are provided for in vitro and in vivo use.
  • Low-nanomolar IRAK4 potency (IC50 2.8 nM).
  • Also inhibits cGAS (IC50 2.1 nM).
  • High purity (99.55%).
  • Soluble in DMSO (25 mg/mL); in vivo formulation protocols provided.
  • Light yellow solid with recommended storage conditions for powder and solutions.