Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IRAK4-IN-4 25mg | 1850276-58-2 | 25 MG
SDP

Catalog No. 5000229398
Encompass_Preferred

IRAK4-IN-4 is a research-grade small-molecule inhibitor of interleukin-1 receptor-associated kinase 4 (IRAK4) with demonstrated potency in enzymatic assays.

  • Potent IRAK4 inhibitor (IC50 = 2.8 nM).
  • Inhibits cyclic GMP-AMP synthase (cGAS) with IC50 = 2.1 nM.
  • High purity (99.55%).
  • Molecular formula C22H16N2O2; molecular weight 340.37.
  • Light yellow to yellow solid supplied in milligram quantities.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).

Catalog No. 50-002-29398 Supplier Medchemexpress LLC Supplier No. HY11418125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IRAK4-IN-4 is a research-grade small-molecule inhibitor of interleukin-1 receptor-associated kinase 4 (IRAK4) with demonstrated potency in enzymatic assays.

  • Potent IRAK4 inhibitor (IC50 = 2.8 nM).
  • Inhibits cyclic GMP-AMP synthase (cGAS) with IC50 = 2.1 nM.
  • High purity (99.55%).
  • Molecular formula C22H16N2O2; molecular weight 340.37.
  • Light yellow to yellow solid supplied in milligram quantities.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.