Learn More
Medchemexpress LLC IRAK4-IN-4 50mg | 1850276-58-2 | 50 MG
SDP
Catalog No. 5000227064
Encompass_Preferred
IRAK4-IN-4 is a research small-molecule inhibitor of interleukin-1 receptor-associated kinase 4 (IRAK4) with additional activity against cyclic GMP-AMP synthase (cGAS). Supplied as a light yellow solid with reported high purity, it is used in biochemical and cellular studies to probe IRAK4- and cGAS-mediated signaling pathways in inflammation and autoimmune disease research.
  • Potent IRAK4 inhibition (IC50 2.8 nM).
  • Potent cGAS inhibition (IC50 2.1 nM).
  • High purity (99.6%).
  • Light yellow solid physical form.
  • Molecular weight 340.37 g/mol.
  • Suitable for preclinical biochemical and cellular assays.
  • Available in multiple small milligram pack sizes for research use.
Catalog No. 50-002-27064 Supplier Medchemexpress LLC Supplier No. HY11418150MG
IRAK4-IN-4 is a research small-molecule inhibitor of interleukin-1 receptor-associated kinase 4 (IRAK4) with additional activity against cyclic GMP-AMP synthase (cGAS). Supplied as a light yellow solid with reported high purity, it is used in biochemical and cellular studies to probe IRAK4- and cGAS-mediated signaling pathways in inflammation and autoimmune disease research.
  • Potent IRAK4 inhibition (IC50 2.8 nM).
  • Potent cGAS inhibition (IC50 2.1 nM).
  • High purity (99.6%).
  • Light yellow solid physical form.
  • Molecular weight 340.37 g/mol.
  • Suitable for preclinical biochemical and cellular assays.
  • Available in multiple small milligram pack sizes for research use.