Learn More
Medchemexpress LLC IRAK4-IN-4 | 5MG
SDP
Catalog No. 5000228845
Encompass_Preferred
IRAK4-IN-4 is a research-grade small-molecule inhibitor of interleukin-1 receptor-associated kinase 4 (IRAK4) that also inhibits cyclic GMP-AMP synthase (cGAS). It demonstrates potent biochemical activity (IRAK4 IC50 = 2.8 nM; cGAS IC50 = 2.1 nM) and is intended for preclinical research into innate immunity and inflammatory pathways. Chemical formula C22H16N2O2; molecular weight 340.37 g/mol.
  • Potent IRAK4 inhibition (IC50 = 2.8 nM).
  • Also inhibits cGAS (IC50 = 2.1 nM).
  • Chemical formula C22H16N2O2 and molecular weight 340.37 g/mol.
  • High purity, approx. 99.6%.
  • Suitable for in vitro and preclinical research applications.
Catalog No. 50-002-28845 Supplier Medchemexpress LLC Supplier No. HY1141815MG
IRAK4-IN-4 is a research-grade small-molecule inhibitor of interleukin-1 receptor-associated kinase 4 (IRAK4) that also inhibits cyclic GMP-AMP synthase (cGAS). It demonstrates potent biochemical activity (IRAK4 IC50 = 2.8 nM; cGAS IC50 = 2.1 nM) and is intended for preclinical research into innate immunity and inflammatory pathways. Chemical formula C22H16N2O2; molecular weight 340.37 g/mol.
  • Potent IRAK4 inhibition (IC50 = 2.8 nM).
  • Also inhibits cGAS (IC50 = 2.1 nM).
  • Chemical formula C22H16N2O2 and molecular weight 340.37 g/mol.
  • High purity, approx. 99.6%.
  • Suitable for in vitro and preclinical research applications.