Learn More
Medchemexpress LLC IRAK4-IN-6 | 2454244-02-9 | 99.3% | 100 MG
SDP
Catalog No. 5000338374
Encompass_Preferred
IRAK4-IN-6 is an orally efficacious and selective inhibitor of IRAK4 (Interleukin-1 receptor associated kinase 4) with an IC50 of 4 nM. It targets MyD88 L265P mutant diffuse large B cell lymphoma. This product is intended for research use only and is not for sale to patients.
  • Orally efficacious and selective IRAK4 inhibitor
  • IC50 of 4 nM
  • Targets MyD88 L265P mutant diffuse large B cell lymphoma
  • For research use only
Catalog No. 50-003-38374 Supplier Medchemexpress LLC Supplier No. HY130253100MG
IRAK4-IN-6 is an orally efficacious and selective inhibitor of IRAK4 (Interleukin-1 receptor associated kinase 4) with an IC50 of 4 nM. It targets MyD88 L265P mutant diffuse large B cell lymphoma. This product is intended for research use only and is not for sale to patients.
  • Orally efficacious and selective IRAK4 inhibitor
  • IC50 of 4 nM
  • Targets MyD88 L265P mutant diffuse large B cell lymphoma
  • For research use only