Learn More
Cayman Chemical IRAK4 Inhibitor 1, Purity: ≥98%, CAS: 1820787-94-7, 1 mg
SDP
Catalog No. 502918177
Encompass_Preferred
An IRAK4 inhibitor (IC50 = 7 nM); reduces resiquimod-induced increases in TNF-α levels in isolated human PBMCs (EC50 = 300 nM)
Catalog No. 50-291-8177 Supplier Cayman Chemical Supplier No. 375481 MG
An IRAK4 inhibitor (IC50 = 7 nM); reduces resiquimod-induced increases in TNF-α levels in isolated human PBMCs (EC50 = 300 nM)