Learn More
Cayman Chemical IRAK4humnrecombInt 100ug
SDP
Catalog No. 502943389
Encompass_Preferred
Source: Active recombinant human IRAK4 expressed in insect cells • Amino acids: 154-460 • MW: 34.4 kDa
Catalog No. 50-294-3389 Supplier Cayman Chemical Supplier No. 42144100 UG
Source: Active recombinant human IRAK4 expressed in insect cells • Amino acids: 154-460 • MW: 34.4 kDa