Learn More
TARGETMOL CHEMICALS INC Irbesartan 100MG
SDP
Catalog No. 502877890
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Irbesartan (SR-47436) is an Angiotensin 2 Receptor Blocker whose mechanism of action involves antagonizing the Angiotensin 2 Receptor. Purity 99.92%
Catalog No. 50-287-7890 Supplier TARGETMOL CHEMICALS INC Supplier No. T1615100MG
Add to Cart
Add to Cart
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Irbesartan (SR-47436) is an Angiotensin 2 Receptor Blocker whose mechanism of action involves antagonizing the Angiotensin 2 Receptor. Purity 99.92%