Learn More
Medchemexpress LLC Irbesartan | 138402-11-6 | 100.0% | 5 MG
SDP
Catalog No. 5000407340
Encompass_Preferred
Irbesartan is an orally active Angiotensin II type 1 (AT1) receptor blocker (ARB). It functions by relaxing blood vessels, which aids in lowering blood pressure and enhancing the supply of blood and oxygen to the heart. This compound is utilized for research studies concerning high blood pressure, heart failure, and diabetic kidney disease.
  • Functions as an orally active Ang II type 1 (AT1) receptor blocker
  • Relaxes blood vessels to lower blood pressure and increase blood and oxygen supply to the heart
  • Utilized in research for high blood pressure, heart failure, and diabetic kidney disease
  • Reduces Th22 cell chemotaxis
  • Suppresses Th22 cell differentiation
  • Inhibits Th22 cells related proinflammatory response of TECs
  • Exerts renoprotective effects
  • Relieves systemic inflammation and renal fibrosis in hypertension models
Catalog No. 50-004-07340 Supplier Medchemexpress LLC Supplier No. HYB02025MG
Irbesartan is an orally active Angiotensin II type 1 (AT1) receptor blocker (ARB). It functions by relaxing blood vessels, which aids in lowering blood pressure and enhancing the supply of blood and oxygen to the heart. This compound is utilized for research studies concerning high blood pressure, heart failure, and diabetic kidney disease.
  • Functions as an orally active Ang II type 1 (AT1) receptor blocker
  • Relaxes blood vessels to lower blood pressure and increase blood and oxygen supply to the heart
  • Utilized in research for high blood pressure, heart failure, and diabetic kidney disease
  • Reduces Th22 cell chemotaxis
  • Suppresses Th22 cell differentiation
  • Inhibits Th22 cells related proinflammatory response of TECs
  • Exerts renoprotective effects
  • Relieves systemic inflammation and renal fibrosis in hypertension models