Learn More
Medchemexpress LLC Irbesartan | 138402-11-6 | 99.9% | 1 ML
SDP
Catalog No. 5000407424
Encompass_Preferred
Irbesartan (SR-47436) is an orally active Angiotensin II type 1 (AT1) receptor blocker. It helps relax blood vessels, lower blood pressure, and increase blood and oxygen supply to the heart. It is used in research for high blood pressure, heart failure, and diabetic kidney disease.
  • Inhibits Angiotensin II-induced proliferation of human aortic smooth muscle cells.
  • Inhibits human MATE1-mediated ASP+ uptake in HEK293 cells.
  • Acts as an antagonist to Angiotensin II, inhibiting its increase in cytosolic calcium concentration.
  • Reduces and suppresses Th22 cells chemotaxis and differentiation.
  • Inhibits Th22 cells related proinflammatory responses in TECs.
Catalog No. 50-004-07424 Supplier Medchemexpress LLC Supplier No. HYB020210MM/1ML
Add to Cart
Add to Cart
Irbesartan (SR-47436) is an orally active Angiotensin II type 1 (AT1) receptor blocker. It helps relax blood vessels, lower blood pressure, and increase blood and oxygen supply to the heart. It is used in research for high blood pressure, heart failure, and diabetic kidney disease.
  • Inhibits Angiotensin II-induced proliferation of human aortic smooth muscle cells.
  • Inhibits human MATE1-mediated ASP+ uptake in HEK293 cells.
  • Acts as an antagonist to Angiotensin II, inhibiting its increase in cytosolic calcium concentration.
  • Reduces and suppresses Th22 cells chemotaxis and differentiation.
  • Inhibits Th22 cells related proinflammatory responses in TECs.