Learn More
Medchemexpress LLC Irbesartan | 138402-11-6 | 99.95% | 500 MG
SDP
Catalog No. 5000405765
Encompass_Preferred
Irbesartan (SR-47436) is an orally active Ang II type 1 (AT1) receptor blocker (ARB) that can relax blood vessels, lower blood pressure, and increase the supply of blood and oxygen to the heart. It is used for the research of high blood pressure, heart failure, and diabetic kidney disease. This product is for research use only and not for sale to patients.
  • Relaxes blood vessels
  • Lowers blood pressure
  • Increases blood and oxygen supply to the heart
  • Used in research for high blood pressure
  • Used in research for heart failure
  • Used in research for diabetic kidney disease
Catalog No. 50-004-05765 Supplier Medchemexpress LLC Supplier No. HYB0202500MG
Irbesartan (SR-47436) is an orally active Ang II type 1 (AT1) receptor blocker (ARB) that can relax blood vessels, lower blood pressure, and increase the supply of blood and oxygen to the heart. It is used for the research of high blood pressure, heart failure, and diabetic kidney disease. This product is for research use only and not for sale to patients.
  • Relaxes blood vessels
  • Lowers blood pressure
  • Increases blood and oxygen supply to the heart
  • Used in research for high blood pressure
  • Used in research for heart failure
  • Used in research for diabetic kidney disease