Learn More
Apexbio Technology LLC Irbesartan 138402-11-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061826
Encompass_Preferred
Irbesartan (CAS 138402-11-6) is a small molecule antagonist of the angiotensin II type 1 (AT1) receptor By selectively blocking AT1 receptor activity irbesartan inhibits angiotensin II-induced vasoconstriction thereby promoting vasodilation and reducing blood pressure In biomedical research irbesartan serves as an important pharmacological tool to investigate cardiovascular regulation hypertension pathogenesis and related signaling pathways Additionally its use extends to studies exploring therapeutic strategies for cardiovascular disorders
Catalog No. 50-000-61826 Supplier Apexbio Technology LLC Supplier No. A59705.1
Irbesartan (CAS 138402-11-6) is a small molecule antagonist of the angiotensin II type 1 (AT1) receptor By selectively blocking AT1 receptor activity irbesartan inhibits angiotensin II-induced vasoconstriction thereby promoting vasodilation and reducing blood pressure In biomedical research irbesartan serves as an important pharmacological tool to investigate cardiovascular regulation hypertension pathogenesis and related signaling pathways Additionally its use extends to studies exploring therapeutic strategies for cardiovascular disorders