Learn More
Medchemexpress LLC Irbesartan | 200 MG
SDP
Catalog No. 5000407337
Encompass_Preferred
Irbesartan (SR-47436) is an orally active Ang II type 1 (AT1) receptor blocker (ARB). It functions by relaxing the blood vessels, which in turn helps to lower blood pressure and increase the supply of blood and oxygen to the heart. This compound can be utilized for research purposes focusing on conditions such as high blood pressure, heart failure, and diabetic kidney disease.
  • Orally active Ang II type 1 (AT1) receptor blocker (ARB)
  • Relaxes blood vessels
  • Lowers blood pressure
  • Increases blood and oxygen supply to the heart
  • Useful for research on high blood pressure
  • Useful for research on heart failure
  • Useful for research on diabetic kidney disease
Catalog No. 50-004-07337 Supplier Medchemexpress LLC Supplier No. HYB0202200MG
Add to Cart
Add to Cart
Irbesartan (SR-47436) is an orally active Ang II type 1 (AT1) receptor blocker (ARB). It functions by relaxing the blood vessels, which in turn helps to lower blood pressure and increase the supply of blood and oxygen to the heart. This compound can be utilized for research purposes focusing on conditions such as high blood pressure, heart failure, and diabetic kidney disease.
  • Orally active Ang II type 1 (AT1) receptor blocker (ARB)
  • Relaxes blood vessels
  • Lowers blood pressure
  • Increases blood and oxygen supply to the heart
  • Useful for research on high blood pressure
  • Useful for research on heart failure
  • Useful for research on diabetic kidney disease