Learn More
Selleck Chemical LLC Irbesartan 50mg 138402-11-6 BMS-186295, SR-47436
SDP
Supplier:  Selleck Chemical LLC S150750MG
Encompass_Preferred
Irbesartan (BMS-186295, SR-47436) is a highly potent and specific angiotensin II type 1 (AT1) receptor antagonist with IC50 of 1.3 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-769-4