Learn More
Cayman Chemical Irbesartan-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1216883-23-6, 1 mg
SDP
Catalog No. 503015697
Encompass_Preferred
An internal standard for the quantification of irbesartan by GC- or LC-MS
Catalog No. 50-301-5697 Supplier Cayman Chemical Supplier No. 255091 MG
An internal standard for the quantification of irbesartan by GC- or LC-MS