Learn More
Cayman Chemical Irbesartn-d4 500ug
SDP
Catalog No. 503015755
Encompass_Preferred
An internal standard for the quantification of irbesartan by GC- or LC-MS
Catalog No. 50-301-5755 Supplier Cayman Chemical Supplier No. 25509500 UG
An internal standard for the quantification of irbesartan by GC- or LC-MS