Learn More
Selleck Chemical LLC Tucatinib (Irbinitinib, ONT-380) 25mg 937263-43-9 ARRY-380
SDP
Catalog No. 501366438
Encompass_Preferred
Tucatinib (Irbinitinib, ONT-380, ARRY-380) is an oral, potent, selective, reversible and ATP-competitive small-molecule inhibitor of ErbB-2 (also called HER2) with IC50s of 8 nM and 7 nM for ErbB-2 and p95 HER2, respectively in cell-based assays, showing ~500-fold selective for HER2 vs EGFR. It has potential antineoplastic activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6438 Supplier Selleck Chemical LLC Supplier No. S836225MG
Add to Cart
Add to Cart
Tucatinib (Irbinitinib, ONT-380, ARRY-380) is an oral, potent, selective, reversible and ATP-competitive small-molecule inhibitor of ErbB-2 (also called HER2) with IC50s of 8 nM and 7 nM for ErbB-2 and p95 HER2, respectively in cell-based assays, showing ~500-fold selective for HER2 vs EGFR. It has potential antineoplastic activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.