Learn More
Medchemexpress LLC HY-P1587 1mg Medchemexpress, IRBP (1-20), human CAS:298202-25-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP15871MG
Encompass_Preferred
Medchemexpress, HY-P1587 1mg IRBP (1-20), human CAS:298202-25-2 IRBP (1-20), human contains a major epitope for the H-2b haplotype. IRBP (1-20), human induces experimental autoimmune uveoretinitis (EAU) in H-2b mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3068