Learn More
Medchemexpress LLC HY-P1587A 1mg Medchemexpress, IRBP (1-20), human (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1587A1MG
Encompass_Preferred
Medchemexpress, HY-P1587A 1mg IRBP (1-20), human (TFA) CAS: IRBP (1-20), human TFA contains a major epitope for the H-2b haplotype. IRBP (1-20), human TFA induces experimental autoimmune uveoretinitis (EAU) in H-2b mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3069