Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IRBP(651-670) human, mouse | 1977546-93-2 | 99.13% | 2107.32 | 5 MG
SDP

Catalog No. 5000422496
Encompass_Preferred

IRBP (651-670) human, mouse is an epitope and uveitis inducer naturally processed from the conserved region of native IRBP. It increases the levels of proinflammatory cytokines in ocular tissues (IL-1β, IL-6, TNFα, IL-17A, and IL-17F). This fragment is conserved among human, mouse, and bovine species, and induces experimental autoimmune uveitis, making it applicable to related research.

  • Epitope and uveitis inducer
  • Increases proinflammatory cytokine levels (IL-1β, IL-6, TNFα, IL-17A, IL-17F)
  • Conserved among human, mouse, and bovine species
  • Induces experimental autoimmune uveitis
  • White to off-white solid
  • Soluble in DMSO (100 mg/mL) and H₂O (2.5 mg/mL)
  • Soluble in PBS (0.5 mg/mL) for in vivo studies
  • Powder storage at -80°C for 2 years or -20°C for 1 year
  • In solvent storage at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-22496 Supplier Medchemexpress LLC Supplier No. HYP19245MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IRBP (651-670) human, mouse is an epitope and uveitis inducer naturally processed from the conserved region of native IRBP. It increases the levels of proinflammatory cytokines in ocular tissues (IL-1β, IL-6, TNFα, IL-17A, and IL-17F). This fragment is conserved among human, mouse, and bovine species, and induces experimental autoimmune uveitis, making it applicable to related research.

  • Epitope and uveitis inducer
  • Increases proinflammatory cytokine levels (IL-1β, IL-6, TNFα, IL-17A, IL-17F)
  • Conserved among human, mouse, and bovine species
  • Induces experimental autoimmune uveitis
  • White to off-white solid
  • Soluble in DMSO (100 mg/mL) and H₂O (2.5 mg/mL)
  • Soluble in PBS (0.5 mg/mL) for in vivo studies
  • Powder storage at -80°C for 2 years or -20°C for 1 year
  • In solvent storage at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.