Learn More
Cayman Chemical IRBP651-670humntrIfl 10mg
SDP
Catalog No. 502915531
Encompass_Preferred
A peptide inducer of experimental autoimmune uveoretinitis; has been used to induce autoimmune uveoretinitis in C57BL/6 mice; immunization with IRBP651-670 increases ocular IL-1β, IL-6, IL-17, TNF-α, and IFN-γ levels, immune cell infiltration, and photoreceptor damage in mice at 300 UG/animal
Catalog No. 50-291-5531 Supplier Cayman Chemical Supplier No. 3695310 MG
A peptide inducer of experimental autoimmune uveoretinitis; has been used to induce autoimmune uveoretinitis in C57BL/6 mice; immunization with IRBP651-670 increases ocular IL-1β, IL-6, IL-17, TNF-α, and IFN-γ levels, immune cell infiltration, and photoreceptor damage in mice at 300 UG/animal