Learn More
LICOR IRDye 680LT Goat anti-Mouse IgM ( chain specific) Secondary Antibody (0.5 mg)
Supplier:  LICOR 92668080
Encompass_Preferred
Immunogen: Mouse IgM paraproteins | Applications: IRDye 680LT Goat anti-Mouse IgM (µ chain specific) Secondary Antibody is recommended for Western Blot, Protein Array, Immunohistochemistry, Microscopy, 2D Gel Detection, Tissue Section Imaging. It is not recommended for Small Animal Imaging, In-Cell Western Assay, and On-Cell Western Assay | Formulation: IRDye 680LT Secondary Antibodies are supplied as purified immunoglobulin conjugates, lyophilized in phosphate-buffered saline, pH 7.4. Protect from light. Store at 4 °C prior to reconstitution. Each vial contains 10 mg/mL BSA (free of IgG and protease) as a stabilizer and 0.01% sodium azide as a preservative, after reconstitution. Concentration is 1.0 mg/mL when reconstituted as directed. Refer to the pack insert for details on reconstitution.
Catalog No. 50-489-551