Learn More
LICOR IRDye 680LT Maleimide Infrared Dye (0.5mg)
Catalog No. NC2216029
Encompass_Preferred
IRDye 680LT Maleimide will label molecules with free sulfhydryl (–SH) groups, such as cysteine residues in proteins. Conjugation can be performed at physiological pH.
This item is not returnable. View return policy
Catalog No. NC2216029 Supplier LICOR Supplier No. 92971008
IRDye 680LT Maleimide will label molecules with free sulfhydryl (–SH) groups, such as cysteine residues in proteins. Conjugation can be performed at physiological pH.