Learn More
LICORbio IRDye 680LT Protein Labeling Kits (microscale kit)
Catalog No. NC2216044
Encompass_Preferred
IRDye 680LT Protein Labeling Kits can be used to label antibodies and other proteins for applications such as Western blots and microscopy. The microscale kit is optimized for labeling 100 μg of protein with molecular weight 14 - 200 kDa. This kit contains 3 x 0.125 mg IRDye 680LT Reactive Dye vials, 0.5 mL 1 M Potassium Phosphate (K2HPO4) (pH 9), 25 mL 1X PBS, 0.5 mL ultra-pure water, 3 x Pierce Zeba Desalting Spin Columns, Pierce Zeba Desalting Spin Column instructions, and Protocol for IRDye 680LT Protein Labeling Kit - Microscale
This item is not returnable. View return policy
Catalog No. NC2216044 Supplier LICORbio Supplier No. 92838070
IRDye 680LT Protein Labeling Kits can be used to label antibodies and other proteins for applications such as Western blots and microscopy. The microscale kit is optimized for labeling 100 μg of protein with molecular weight 14 - 200 kDa. This kit contains 3 x 0.125 mg IRDye 680LT Reactive Dye vials, 0.5 mL 1 M Potassium Phosphate (K2HPO4) (pH 9), 25 mL 1X PBS, 0.5 mL ultra-pure water, 3 x Pierce Zeba Desalting Spin Columns, Pierce Zeba Desalting Spin Column instructions, and Protocol for IRDye 680LT Protein Labeling Kit - Microscale