Learn More
LICORbio IRDye 680LT Protein Labeling Kits (high molecular weight kit)
Catalog No. NC2216042
Encompass_Preferred
IRDye 680LT Protein Labeling Kits can be used to label antibodies and other proteins for applications such as Western blots and microscopy. The high molecular weight kit is optimized for labeling 1.0 mg of protein with molecular weight 45 - 200 kDa. This kit contains 3 x 0.125 mg IRDye 680LT Reactive Dye vials, 0.5 mL 1 M Potassium Phosphate (K2HPO4) (pH 9), 25 mL 1X PBS, 0.5 mL ultra-pure water, 3 x Pierce Zeba™ Desalting Spin Columns, Pierce Zeba Desalting Spin column instructions and Protocol for IRDye 680LT Protein Labeling Kit - High MW
This item is not returnable. View return policy
Catalog No. NC2216042 Supplier LICORbio Supplier No. 92838066
IRDye 680LT Protein Labeling Kits can be used to label antibodies and other proteins for applications such as Western blots and microscopy. The high molecular weight kit is optimized for labeling 1.0 mg of protein with molecular weight 45 - 200 kDa. This kit contains 3 x 0.125 mg IRDye 680LT Reactive Dye vials, 0.5 mL 1 M Potassium Phosphate (K2HPO4) (pH 9), 25 mL 1X PBS, 0.5 mL ultra-pure water, 3 x Pierce Zeba™ Desalting Spin Columns, Pierce Zeba Desalting Spin column instructions and Protocol for IRDye 680LT Protein Labeling Kit - High MW