Learn More
LICORbio IRDye 680RD Alkyne Infrared Dye (5mg)
Catalog No. NC2209471
Encompass_Preferred
IRDye 680RD Alkyne provides the functionality for preferential labeling of molecules containing azide groups (–N3) with IRDye 680RD.
This item is not returnable. View return policy
Catalog No. NC2209471 Supplier LICORbio Supplier No. 92965006
IRDye 680RD Alkyne provides the functionality for preferential labeling of molecules containing azide groups (–N3) with IRDye 680RD.