Learn More
LICORbio IRDye 680RD Azide Infrared Dye (5mg)
Catalog No. NC2209465
Encompass_Preferred
IRDye 680RD Azide provides the functionality for preferential labeling of molecules that contain either the alkyne or dibenzocyclooctyne (DBCO) group.
This item is not returnable. View return policy
Catalog No. NC2209465 Supplier LICORbio Supplier No. 92965005
IRDye 680RD Azide provides the functionality for preferential labeling of molecules that contain either the alkyne or dibenzocyclooctyne (DBCO) group.