Learn More
LICORbio IRDye 680RD Azide Infrared Dye (0.5 mg)
Catalog No. NC0672168
Encompass_Preferred
IRDye 680RD Azide provides the functionality for preferential labeling of molecules that contain either the alkyne or dibenzocyclooctyne (DBCO) group.
Catalog No. NC0672168 Supplier LICORbio Supplier No. 92960005
IRDye 680RD Azide provides the functionality for preferential labeling of molecules that contain either the alkyne or dibenzocyclooctyne (DBCO) group.