Learn More
LICORbio IRDye 680RD Carboxylate (100 nmol)
Catalog No. NC1797550
Encompass_Preferred
Assays that use IRDye 680RD conjugates (such as small animal imaging and cell binding assays) may require a "dye-only" control for potential effects or retention of the dye. Carboxylate (non-reactive) form of IRDye 680RD is an ideal control.
Catalog No. NC1797550 Supplier LICORbio Supplier No. 92908980
Assays that use IRDye 680RD conjugates (such as small animal imaging and cell binding assays) may require a "dye-only" control for potential effects or retention of the dye. Carboxylate (non-reactive) form of IRDye 680RD is an ideal control.