Learn More
LICOR IRDye 680RD Goat anti-Mouse IgM ( chain specific) Secondary Antibody (0.1 mg)
Supplier:  LICOR 92568180
Encompass_Preferred
Immunogen: Mouse lgM paraproteins | Application: IRDye 680RD Goat anti-Mouse IgM (µ chain specific) Secondary Antibody is highly recommended for Western Blot, In-Cell Western™ Assay, On-Cell Western Assay, Protein Array, Immunohistochemistry, Small Animal Imaging, Microscopy, 2D Gel Detection, and Tissue Section Imaging. | Formulation: IRDye 680RD Secondary Antibodies are supplied as purified immunoglobulin conjugates, lyophilized in phosphate-buffered saline, pH 7.4. Protect from light. Store at 4 °C prior to reconstitution. Each vial contains 10 mg/mL BSA (free of IgG and protease) as a stabilizer and 0.01% sodium azide as a preservative, after reconstitution. Concentration is 1.0 mg/mL when reconstituted as directed. Refer to the pack insert for details on reconstitution.
Catalog No. NC2209171