Learn More
LICOR IRDye 680RD Maleimide Infrared Dye (5mg)
Catalog No. NC2216036
Encompass_Preferred
IRDye 680RD Maleimide will label molecules with free sulfhydryl (-SH) groups, such as cysteine residues in proteins. Conjugation can be performed at physiological pH.
This item is not returnable. View return policy
Catalog No. NC2216036 Supplier LICOR Supplier No. 92971051
IRDye 680RD Maleimide will label molecules with free sulfhydryl (-SH) groups, such as cysteine residues in proteins. Conjugation can be performed at physiological pH.