Learn More
LICOR IRDye 680RD NHS Ester (50 mg)
Supplier:  LICOR 92970052
Encompass_Preferred
IRDye infrared dyes from LI-COR Biosciences are available for researchers developing applications using near-infrared (NIR) fluorescence. These unique dyes are used in a variety of NIR imaging applications including Western blotting, plate-based assays, protein arrays, tissue section imaging, and molecular activity measurements. Standard NHS ester chemistry is used to produce custom probes labeled with LI-COR IRDye infrared dyes. The NHS ester reactive group provides the functionality for labeling primary and secondary amines, such as lysine residues in proteins.
Catalog No. NC2209484
Add to Cart
Request a Quote