Learn More
LICOR IRDye 680RD Protein Labeling Kits (high molecular weight kit)
Catalog No. NC1701839
Encompass_Preferred
IRDye 680RD Protein Labeling Kits can be used to label antibodies and other proteins for Western blotting, In-Cell Western™ assays, in vivo imaging, and whole organ or tissue section applications. The high molecular weight kit is optimized for labeling 1.0 mg of protein with molecular weight 45 - 200 kDa. This kit contains, 3 x 0.1 mg IRDye 680RD Reactive Dye vials, 0.5 mL 1 M Potassium Phosphate (K2HPO4) (pH 9), 25 mL 1X PBS, 0.5 mL ultra-pure water, 3 x Pierce Zeba™ Desalting Spin Columns, Pierce Zeba Desalting Spin column instructions, and Protocol for IRDye 680RD Protein Labeling Kit - High MW
Catalog No. NC1701839 Supplier LICOR Supplier No. 92838072
Add to Cart
Add to Cart
IRDye 680RD Protein Labeling Kits can be used to label antibodies and other proteins for Western blotting, In-Cell Western™ assays, in vivo imaging, and whole organ or tissue section applications. The high molecular weight kit is optimized for labeling 1.0 mg of protein with molecular weight 45 - 200 kDa. This kit contains, 3 x 0.1 mg IRDye 680RD Reactive Dye vials, 0.5 mL 1 M Potassium Phosphate (K2HPO4) (pH 9), 25 mL 1X PBS, 0.5 mL ultra-pure water, 3 x Pierce Zeba™ Desalting Spin Columns, Pierce Zeba Desalting Spin column instructions, and Protocol for IRDye 680RD Protein Labeling Kit - High MW